missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypyridine, 98%

Catalog No. AAA1391036
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1391036 500 g
AAA1391018 50 g
AAA1391022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1391036 Supplier Thermo Scientific Chemicals Supplier No. A1391036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-00-2 | C5H5NO | 95.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-00-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00006378
InChI Key GRFNBEZIAWKNCO-UHFFFAOYSA-N
Synonym 3-hydroxypyridine, 3-pyridinol, 3-pyridol, 3-pyridone, 3-oxopyridine, beta-hydroxypyridine, m-hydroxypyridine, 3-pyridyl alcohol, 3-hydoxypyridine, unii-4kbe4p5b6s
PubChem CID 7971
IUPAC Name pyridin-3-ol
SMILES C1=CC(=CN=C1)O

Specifications

Melting Point 125°C to 128°C
Boiling Point 180°C (22 mmHg)
Flash Point 173°C (343°F)
Quantity 500 g
Beilstein 105699
Sensitivity Light sensitive
Formula Weight 95.1
Percent Purity 98%
Chemical Name or Material 3-Hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-637-4
RTECSNumber UU7701400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only