missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypyridine-2-carboxylic acid, 98%
Description
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.
Solubility
Soluble in water (0.25g in 10ml).
Notes
Keep container tightly closed. Store away from oxidizing agents.
3-Hydroxypyridine-2-carboxylic acid is used as a matrix for nucleotides in MALDI mass spectrometry analyses.
Solubility
Soluble in water (0.25g in 10ml).
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | ∽214°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 118954 |
| Solubility Information | Soluble in water (0.25g in 10ml). |
| Formula Weight | 139.11 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Hydroxypyridine-2-carboxylic acid |
Safety and Handling
| Hazard Category | H302+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 212-859-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only