accessibility menu, dialog, popup

UserName

CAS RN 55321-99-8

IUPAC Name: 3-oxo-3,4-dihydropyrazine-2-carboxamide
Synonyms: 3-oxo-4H-pyrazine-2-carboxamide
Molecular Weight (g/mol): 139.11
Molecular Formula: C5H5N3O2
InChi Key: SZPBAPFUXAADQV-UHFFFAOYSA-N
SMILES: NC(=O)C1=NC=CNC1=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)