Learn More
3-Hydroxypyrazine-2-carboxamide, 98%, Thermo Scientific Chemicals
$264.79 - $264.79
Chemical Identifiers
| CAS | 55321-99-8 |
|---|---|
| Molecular Formula | C5H5N3O2 |
| Molecular Weight (g/mol) | 139.114 |
| MDL Number | MFCD00233977 |
| InChI Key | SZPBAPFUXAADQV-UHFFFAOYSA-N |
| Synonym | 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry |
| PubChem CID | 294642 |
| IUPAC Name | 2-oxo-1H-pyrazine-3-carboxamide |
| SMILES | C1=CN=C(C(=O)N1)C(=O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2825006
|
Thermo Scientific Chemicals
H2825006 |
5 g |
Each for $264.79
|
|
|||||
Description
Apart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsApart from biological importance of pyridine-2-carboxamide (picolinamide, pia) as well as of its derivatives such as 3-hydroxypyridine-2-carboxamide (3-OHpia), as ligands they have found their extensive application in coordina- tion chemistry in crystal engineering .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 55321-99-8 | |
| 139.114 | |
| SZPBAPFUXAADQV-UHFFFAOYSA-N | |
| 294642 | |
| C1=CN=C(C(=O)N1)C(=O)N |
| C5H5N3O2 | |
| MFCD00233977 | |
| 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry | |
| 2-oxo-1H-pyrazine-3-carboxamide |
Specifications
| 55321-99-8 | |
| C5H5N3O2 | |
| 5 g | |
| SZPBAPFUXAADQV-UHFFFAOYSA-N | |
| 2-oxo-1H-pyrazine-3-carboxamide | |
| 294642 | |
| 98% |
| ∽270°C (decomposition) | |
| MFCD00233977 | |
| 3-hydroxypyrazine-2-carboxamide, 3-oxo-3,4-dihydropyrazine-2-carboxamide, pyrazinecarboxamide, 3,4-dihydro-3-oxo, 3-hydroxy-2-pyrazinecarboxamide, pyrazine-2-carboxamide, 3-hydroxy, 3-hydroxypyrazine-2-carboxamine, 3-oxo-4h-pyrazine-2-carboxamide, t6mv dnj cvz wln, d09vhw, d0s5ry | |
| C1=CN=C(C(=O)N1)C(=O)N | |
| 139.114 | |
| 139.12 | |
| 3-Hydroxypyrazine-2-carboxamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only