missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionitrile, 97%

CAS: 109-78-4 | C3H5NO | 71.079 g/mol

Supplier:  Thermo Scientific Chemicals L125700B

 View more versions of this product

Catalog No. AAL125700B


Only null left
Explore available promotions

Description

Description

3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Hydroxypropionitrile
-46°C
227°C to 228°C
C3H5NO
HO(CH2)2CN
1000 g
14,3794
Fully miscible in water.
C(CO)C#N
71.079
71.08
109-78-4
1.041
128°C (262°F)
1.425
MFCD00002826
635773
3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
WSGYTJNNHPZFKR-UHFFFAOYSA-N
3-hydroxypropanenitrile
8011
98%

Chemical Identifiers

109-78-4
71.079
WSGYTJNNHPZFKR-UHFFFAOYSA-N
8011
C(CO)C#N
C3H5NO
MFCD00002826
3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
3-hydroxypropanenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 203-704-8

RTECSNumber : MU5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only