missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypropionitrile, 97%

Catalog No. p-7045950
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
Catalog No. Quantity
AAL1257014 25 g
AAL1257030 250 g
AAL125700B 1000 g
3 options

Catalog No. AAL1257014

Supplier: Thermo Scientific Chemicals L1257014

Only null left

CAS: 109-78-4 | C3H5NO | 71.079 g/mol

3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxypropionitrile has been used as polar solvent to investigate the orientation of the phospholipid at the surface of the polar solvents by neutral impact collision ion scattering spectroscopy and as a reagent for carboxyl protection in peptide synthesis. It has been used also in solid state composite electrolyte for dye-sensitized solar cells.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 109-78-4
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.079
MDL Number MFCD00002826
InChI Key WSGYTJNNHPZFKR-UHFFFAOYSA-N
Synonym 3-hydroxypropionitrile, hydracrylonitrile, 2-cyanoethanol, ethylene cyanohydrin, propanenitrile, 3-hydroxy, beta-cyanoethanol, glycol cyanohydrin, methanolacetonitrile, hydroxypropanenitrile, 2-hydroxycyanoethane
PubChem CID 8011
IUPAC Name 3-hydroxypropanenitrile
SMILES C(CO)C#N

Specifications

Melting Point -46°C
Density 1.041
Boiling Point 227°C to 228°C
Flash Point 128°C (262°F)
Refractive Index 1.425
Linear Formula HO(CH2)2CN
Quantity 25 g
Beilstein 635773
Merck Index 14,3794
Solubility Information Fully miscible in water.
Formula Weight 71.08
Percent Purity 98%
Chemical Name or Material 3-Hydroxypropionitrile
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 203-704-8
RTECSNumber MU5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only