missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypropionic Acid (contains varying amounts of 3,3'-Oxydipropionic Acid), TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 10 g |
| Formula Weight | 90.08 |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Hydroxypropionic Acid (contains varying amounts of 3,3′-Oxydipropionic Acid) (ca. 30% in Wat |
Safety and Handling
| EINECSNumber | (2)-1369 |
| RTECSNumber | MU4375000 |
| TSCA | No |
| Recommended Storage | Refrigerator |