accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 503-66-2

IUPAC Name: 3-hydroxypropanoic acid
Synonyms: 3-hydroxy-propanoic acid hydroxy acrylic acid β-lactic acid
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: ALRHLSYJTWAHJZ-UHFFFAOYSA-N
SMILES: OCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results