missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypiperidine 98.0+%, TCI America™
$52.19 - $164.03
Chemical Identifiers
| CAS | 6859-99-0 |
|---|---|
| Molecular Formula | C5H11NO |
| Molecular Weight (g/mol) | 101.149 |
| MDL Number | MFCD00014591 |
| InChI Key | BIWOSRSKDCZIFM-UHFFFAOYSA-N |
| Synonym | 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine |
| PubChem CID | 23293 |
| IUPAC Name | piperidin-3-ol |
| SMILES | C1CC(CNC1)O |
Chemical Identifiers
| 6859-99-0 | |
| 101.149 | |
| BIWOSRSKDCZIFM-UHFFFAOYSA-N | |
| 23293 | |
| C1CC(CNC1)O |
| C5H11NO | |
| MFCD00014591 | |
| 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine | |
| piperidin-3-ol |
Specifications
| 6859-99-0 | |
| White-Yellow | |
| MFCD00014591 | |
| 1759 | |
| BIWOSRSKDCZIFM-UHFFFAOYSA-N | |
| piperidin-3-ol | |
| 23293 | |
| ≥98.0% (GC) | |
| 3-Hydroxypiperidine |
| 62°C | |
| C5H11NO | |
| 5 g | |
| 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine | |
| C1CC(CNC1)O | |
| 101.149 | |
| 101.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No