missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypiperidine 98.0+%, TCI America™
Supplier: TCI America H041225G
Specifications
| 3-Hydroxypiperidine | |
| 62°C | |
| C5H11NO | |
| 25 g | |
| 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine | |
| C1CC(CNC1)O | |
| 101.149 | |
| 101.15 | |
| Crystalline Powder |
| 6859-99-0 | |
| White-Yellow | |
| MFCD00014591 | |
| 1759 | |
| BIWOSRSKDCZIFM-UHFFFAOYSA-N | |
| piperidin-3-ol | |
| 23293 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6859-99-0 | |
| 101.149 | |
| BIWOSRSKDCZIFM-UHFFFAOYSA-N | |
| 23293 | |
| C1CC(CNC1)O |
| C5H11NO | |
| MFCD00014591 | |
| 3-hydroxypiperidine, 3-piperidinol, 3-hydroxypiperadine, 3-hydroxy piperidine, +/--3-hydroxypiperidine, racemic-3r-piperidin-3-ol, 3-s-hydroxypiperidine, piperidine-3-ol, ppiperidin-3-ol, 3-hydroxy-piperdine | |
| piperidin-3-ol |
Safety and Handling
TSCA : No