missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypicolinic acid matrix substance for MALDI-MS, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015721
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015721 250 mg
111015722 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015721 Supplier Merck Emd Millipore Supplier No. 56197250MG

Specifications

CAS 874-24-8
Linear Formula C6H5 NO3
MDL Number MFCD00006294
Melting Point 213°C to 218°C
Synonym 3-HPA; 3-Hydroxypyridine-2-carboxylic acid
Type Organic Standard
Quantity 250 mg
Formula Weight 139.11
Percent Purity ≥99% (HPLC)
Grade Matrix Substance for MALDI-MS