missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxypicolinic acid, 98%

Catalog No. p-2690399
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC241050050 5 g Glass bottle
AC241050250 25 g Glass bottle
2 options

Catalog No. AC241050050

Supplier: Thermo Scientific Chemicals 241050050

Only null left

CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874-24-8
Molecular Formula C6H4NO3
Molecular Weight (g/mol) 138.10
MDL Number MFCD00006294
InChI Key BRARRAHGNDUELT-UHFFFAOYSA-M
Synonym 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
PubChem CID 13401
ChEBI CHEBI:64342
SMILES OC1=CC=CN=C1C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 208°C to 212°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, V,5, 87
IUPAC Name 3-hydroxypyridine-2-carboxylate
Formula Weight 139.11
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material 3-Hydroxypicolinic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-859-

RUO – Research Use Only