missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymyristic Acid 98.0+%, TCI America™

Supplier:  TCI America H02741G

 View more versions of this product

Catalog No. H02741G


Only null left

Description

Description

Specifications

Specifications

3-Hydroxymyristic Acid
80°C
C14H28O3
1 g
ATRNZOYKSNPPBF-UHFFFAOYSA-N
3-hydroxytetradecanoic acid
16064
244.38
Crystalline Powder
1961-72-4
White
MFCD00059633
3-Hydroxytetradecanoic Acid
CCCCCCCCCCCC(CC(=O)O)O
244.375
CHEBI:85148
≥98.0% (GC,T)

Chemical Identifiers

1961-72-4
244.375
ATRNZOYKSNPPBF-UHFFFAOYSA-N
16064
3-hydroxytetradecanoic acid
C14H28O3
MFCD00059633
3-Hydroxytetradecanoic Acid
CHEBI:85148
CCCCCCCCCCCC(CC(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1340

RTECSNumber : XB8585000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions