accessibility menu, dialog, popup

UserName

H3COHOHO

CAS RN 1961-72-4

IUPAC Name: 3-hydroxytetradecanoic acid
Synonyms: 3-hydroxy-myristic acid
Molecular Weight (g/mol): 244.38
Molecular Formula: C14H28O3
InChi Key: ATRNZOYKSNPPBF-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCC(O)CC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results