missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxymyristic Acid 98.0+%, TCI America™

Catalog No. H02745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02745G 5 g
H02741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02745G Supplier TCI America Supplier No. H02745G

Chemical Identifiers

CAS 1961-72-4
Molecular Formula C14H28O3
Molecular Weight (g/mol) 244.375
MDL Number MFCD00059633
InChI Key ATRNZOYKSNPPBF-UHFFFAOYSA-N
Synonym 3-Hydroxytetradecanoic Acid
PubChem CID 16064
ChEBI CHEBI:85148
IUPAC Name 3-hydroxytetradecanoic acid
SMILES CCCCCCCCCCCC(CC(=O)O)O

Specifications

Melting Point 80°C
Color White
Quantity 5 g
Formula Weight 244.38
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxymyristic Acid
EINECSNumber (2)-1340
RTECSNumber XB8585000
TSCA No