missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)-1-adamantanol 98.0+%, TCI America™

Catalog No. p-7131031
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14151G 1 g
H14155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14151G Supplier TCI America Supplier No. H14151G

Chemical Identifiers

CAS 38584-37-1
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD03105096
InChI Key FORAJDRXEYKDFJ-UHFFFAOYSA-N
Synonym 3-hydroxymethyl adamantan-1-ol, 3-hydroxymethyl-1-adamantanol, tricyclo 3.3.1.13,7 decane-1-methanol, 3-hydroxy, 3-hydroxymethyl-1-adamantol, 3-hydroxy-1-adamantanemethanol, 3-hydroxyadamantyl methanol, 1-hydroxymethyl-3-adamantanol, 3-hydroxy-1-adamantane methanol, 1-hydroxy-3-hydroxymethyladamantane
PubChem CID 12070475
IUPAC Name 3-(hydroxymethyl)adamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)CO

Specifications

Melting Point 158°C
Color White
Quantity 1 g
Formula Weight 182.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Hydroxymethyl)-1-adamantanol