accessibility menu, dialog, popup

UserName

OHOH

CAS RN 38584-37-1

IUPAC Name: 3-(hydroxymethyl)adamantan-1-ol
Synonyms: 3-(hydroxymethyl)adamantan-1-ol
Molecular Weight (g/mol): 182.26
Molecular Formula: C11H18O2
InChi Key: FORAJDRXEYKDFJ-UHFFFAOYNA-N
SMILES: OCC12CC3CC(CC(O)(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results