missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyglutaric acid, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015013
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015013 10 mg
111015014 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015013 Supplier Merck Emd Millipore Supplier No. 0472510MG

Specifications

CAS 638-18-6
Linear Formula C5H8O5
MDL Number MFCD06208235
Synonym beta-Hydroxyglutaric acid; 3-Hydroxypentanedioic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 148.11
Percent Purity ≥95% (GC)
Grade Analytical Standard