missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxycoumarin 98.0+%, TCI America™

Catalog No. H15855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15855G 5 g
H15851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15855G Supplier TCI America Supplier No. H15855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 939-19-5
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00017490
InChI Key MJKVTPMWOKAVMS-UHFFFAOYSA-N
PubChem CID 13650
IUPAC Name 3-hydroxy-2H-chromen-2-one
SMILES OC1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxycoumarin
RTECSNumber GN6797000
TSCA No