accessibility menu, dialog, popup

UserName

CAS RN 939-19-5

IUPAC Name: 3-hydroxy-2H-chromen-2-one
Synonyms: 3 hydroxycoumarin 3-hydroxy-2-benzopyrone
Molecular Weight (g/mol): 162.14
Molecular Formula: C9H6O3
InChi Key: MJKVTPMWOKAVMS-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=CC=C2OC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results