Learn More
3-Hydroxybenzylamine, 96%, Thermo Scientific Chemicals
$308.93 - $905.60
Chemical Identifiers
| CAS | 73604-31-6 |
|---|---|
| Molecular Formula | C7H9NO |
| Molecular Weight (g/mol) | 123.16 |
| MDL Number | MFCD00798977 |
| InChI Key | JNZYADHPGVZMQK-UHFFFAOYSA-N |
| Synonym | 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine |
| PubChem CID | 735894 |
| IUPAC Name | 3-(aminomethyl)phenol |
| SMILES | NCC1=CC(O)=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3570803
|
Thermo Scientific Chemicals
H3570803 |
1 g |
Each for $308.93
|
|
|||||
|
AAH3570806
|
Thermo Scientific Chemicals
H3570806 |
5 g |
Each for $905.60
|
|
|||||
Description
3-Hydroxybenzylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzylamine is used as pharmaceutical intermediate.
Solubility
Soluble in water and ethanol.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
| 73604-31-6 | |
| 123.16 | |
| JNZYADHPGVZMQK-UHFFFAOYSA-N | |
| 735894 | |
| NCC1=CC(O)=CC=C1 |
| C7H9NO | |
| MFCD00798977 | |
| 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine | |
| 3-(aminomethyl)phenol |
Specifications
| 73604-31-6 | |
| C7H9NO | |
| 1 g | |
| Air sensitive | |
| Soluble in water and ethanol. | |
| NCC1=CC(O)=CC=C1 | |
| 123.16 | |
| 123.15 | |
| 3-Hydroxybenzylamine |
| 170°C to 173°C | |
| MFCD00798977 | |
| UN3259 | |
| 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine | |
| JNZYADHPGVZMQK-UHFFFAOYSA-N | |
| 3-(aminomethyl)phenol | |
| 735894 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only