missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzylamine, 96%, Thermo Scientific Chemicals

Catalog No. AAH3570803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3570803 1 g
AAH3570806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3570803 Supplier Thermo Scientific Chemicals Supplier No. H3570803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

3-Hydroxybenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzylamine is used as pharmaceutical intermediate.

Solubility
Soluble in water and ethanol.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 73604-31-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00798977
InChI Key JNZYADHPGVZMQK-UHFFFAOYSA-N
Synonym 3-aminomethyl phenol, 3-hydroxybenzylamine, phenol, 3-aminomethyl, 3-hydroxybenzylamine, 3-hydroxyphenyl methylamine, 3-aminomethyl-phenol, 3-hydroxy-benzylamine, 3-hydroxyphenyl methylamine
PubChem CID 735894
IUPAC Name 3-(aminomethyl)phenol
SMILES NCC1=CC(O)=CC=C1

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
UN Number UN3259
Sensitivity Air sensitive
Solubility Information Soluble in water and ethanol.
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 3-Hydroxybenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only