missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzyl Alcohol 99.0+%, TCI America™

Catalog No. p-7130806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09185G 5 g
H091825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09185G Supplier TCI America Supplier No. H09185G
Only null left

Chemical Identifiers

CAS 620-24-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004643
InChI Key OKVJCVWFVRATSG-UHFFFAOYSA-N
Synonym 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol
PubChem CID 102
ChEBI CHEBI:17069
IUPAC Name 3-(hydroxymethyl)phenol
SMILES C1=CC(=CC(=C1)O)CO

Specifications

Melting Point 72°C
Color White
Boiling Point 123°C
Quantity 5 g
Formula Weight 124.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzyl Alcohol
EINECSNumber (3)-1052
RTECSNumber DA4796700
TSCA No