missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzyl Alcohol, 97%

Catalog No. p-3736590
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC121020050 5 g Glass bottle
1 options

Catalog No. AC121020050

Supplier: Thermo Scientific Chemicals 121020050

Only null left

CAS: 620-24-6 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-24-6
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004643
InChI Key OKVJCVWFVRATSG-UHFFFAOYSA-N
Synonym 3-hydroxybenzyl alcohol, 3-hydroxymethyl phenol, benzenemethanol, 3-hydroxy, 3-methylolphenol, m-hydroxybenzyl alcohol, 3-hydroxybenzenemethanol, benzyl alcohol, m-hydroxy, 3-hydroxybenzylalcohol, 3-oh-benzyl-alcohol, 3-hydroxymethyl-phenol
PubChem CID 102
ChEBI CHEBI:17069
IUPAC Name 3-(hydroxymethyl)phenol
SMILES C1=CC(=CC(=C1)O)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 67°C to 71°C
Color Beige to Brown or Pink
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H4CH2OH
Quantity 5 g
Beilstein 06, 896
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in chloroform,soluble in ethanol and ether
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 210-633-6

RUO – Research Use Only