missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7136283
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043625G 25 g
T0436250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T043625G Supplier TCI America Supplier No. T043625G
Only null left

Chemical Identifiers

CAS 98-17-9
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002299
InChI Key UGEJOEBBMPOJMT-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
PubChem CID 7376
IUPAC Name 3-(trifluoromethyl)phenol
SMILES C1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 178°C
Quantity 25 g
Formula Weight 162.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxybenzotrifluoride
RTECSNumber GP3510000
TSCA Yes