missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzophenone, 98+%

Catalog No. AAA1806103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1806103 1 g
AAA1806106 5 g
AAA1806114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1806103 Supplier Thermo Scientific Chemicals Supplier No. A1806103

CAS: 13020-57-0 | C13H10O2 | 198.221 g/mol

3-Hydroxybenzophenone is a benzophenone metabolite with potential estrogenic and anti-androgenic activity. is a more attractive hydrogen bonding co-material for MCPBA, as the 3-hydroxyl group is unable to form an intramolecular hydrogen bond. It is commonly used as a radical and cationic photo initiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzophenone is a benzophenone metabolite with potential estrogenic and anti-androgenic activity. is a more attractive hydrogen bonding co-material for MCPBA, as the 3-hydroxyl group is unable to form an intramolecular hydrogen bond. It is commonly used as a radical and cationic photo initiators.

Solubility
Soluble in Chloroform, DCM. Ethyl Acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13020-57-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002297
InChI Key SHULEACXTONYPS-UHFFFAOYSA-N
Synonym 3-hydroxybenzophenone, 3-hydroxyphenyl phenyl methanone, m-hydroxybenzophenone, methanone, 3-hydroxyphenyl phenyl, 3-hydroxy-benzophenon, dsstox_cid_27856, dsstox_rid_82609, dsstox_gsid_47880, 3-hydroxyphenyl phenyl ketone
PubChem CID 83050
IUPAC Name (3-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O

Specifications

Melting Point 113°C to 115°C
Quantity 1 g
Solubility Information Soluble in Chloroform,DCM. Ethyl Acetate.
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-879-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only