missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzophenone 98.0+%, TCI America™

$264.70 - $264.70

Chemical Identifiers

CAS 13020-57-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002297
InChI Key SHULEACXTONYPS-UHFFFAOYSA-N
Synonym 3-hydroxybenzophenone, 3-hydroxyphenyl phenyl methanone, m-hydroxybenzophenone, methanone, 3-hydroxyphenyl phenyl, 3-hydroxy-benzophenon, dsstox_cid_27856, dsstox_rid_82609, dsstox_gsid_47880, 3-hydroxyphenyl phenyl ketone
PubChem CID 83050
IUPAC Name (3-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15185G
View Documents
TCI America
H15185G
5 g
Each for $264.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13020-57-0
198.221
SHULEACXTONYPS-UHFFFAOYSA-N
83050
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O
C13H10O2
MFCD00002297
3-hydroxybenzophenone, 3-hydroxyphenyl phenyl methanone, m-hydroxybenzophenone, methanone, 3-hydroxyphenyl phenyl, 3-hydroxy-benzophenon, dsstox_cid_27856, dsstox_rid_82609, dsstox_gsid_47880, 3-hydroxyphenyl phenyl ketone
(3-hydroxyphenyl)-phenylmethanone

Specifications

13020-57-0
C13H10O2
5 g
SHULEACXTONYPS-UHFFFAOYSA-N
(3-hydroxyphenyl)-phenylmethanone
83050
≥98.0% (GC)
3-Hydroxybenzophenone
117°C
MFCD00002297
3-hydroxybenzophenone, 3-hydroxyphenyl phenyl methanone, m-hydroxybenzophenone, methanone, 3-hydroxyphenyl phenyl, 3-hydroxy-benzophenon, dsstox_cid_27856, dsstox_rid_82609, dsstox_gsid_47880, 3-hydroxyphenyl phenyl ketone
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O
198.221
198.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : PC4959730

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'