missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. h0205500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
H0205500G 500 g
H020525G 25 g
H0205100G 100 g
3 options

Catalog No. H0205500G

Supplier: TCI America H0205500G

Only null left

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 203°C
Color White
Quantity 500 g
Formula Weight 138.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzoic Acid
EINECSNumber (3)-1640
RTECSNumber DH1924980
TSCA Yes