missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. H020525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H020525G 25 g
H0205100G 100 g
H0205500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H020525G Supplier TCI America Supplier No. H020525G

Chemical Identifiers

CAS 99-06-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002506
InChI Key IJFXRHURBJZNAO-UHFFFAOYSA-N
Synonym m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova
PubChem CID 7420
ChEBI CHEBI:30764
IUPAC Name 3-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 203°C
Color White
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzoic Acid
EINECSNumber (3)-1640
RTECSNumber DH1924980
TSCA Yes