Learn More
3-Hydroxybenzoic acid, 99%
CAS: 99-06-9 | C7H6O3 | 138.12 g/mol
Supplier: Thermo Scientific Chemicals A1362822
Description
3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzoic acid is used in perfumery. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 3-Hydroxybenzoic acid | |
| 201°C to 203°C | |
| Odorless | |
| MFCD00002506 | |
| 508160 | |
| Slightly soluble in water. | |
| OC(=O)C1=CC=CC(O)=C1 | |
| 138.12 | |
| CHEBI:30764 | |
| 99% |
| 99-06-9 | |
| 220°C (428°F) | |
| C7H6O3 | |
| 100 g | |
| m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova | |
| IJFXRHURBJZNAO-UHFFFAOYSA-N | |
| 3-hydroxybenzoic acid | |
| 7420 | |
| 138.12 |
Chemical Identifiers
| 99-06-9 | |
| 138.12 | |
| IJFXRHURBJZNAO-UHFFFAOYSA-N | |
| 7420 | |
| 3-hydroxybenzoic acid |
| C7H6O3 | |
| MFCD00002506 | |
| m-hydroxybenzoic acid, m-salicylic acid, 3-carboxyphenol, benzoic acid, 3-hydroxy, m-hba, 3-hydroxybenzoate, benzoic acid, m-hydroxy, acido m-idrossibenzoico, meta-hydroxybenzoic acid, kyselina 3-hydroxybenzoova | |
| CHEBI:30764 | |
| OC(=O)C1=CC=CC(O)=C1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 202-726-5
RTECSNumber : DH1924980
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only