missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Hydroxyacetophenone 98.0+%, TCI America™
$61.61 - $521.72
Chemical Identifiers
| CAS | 121-71-1 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00002298 |
| InChI Key | LUJMEECXHPYQOF-UHFFFAOYSA-N |
| Synonym | 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol |
| PubChem CID | 8487 |
| IUPAC Name | 1-(3-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC=C1)O |
Chemical Identifiers
| 121-71-1 | |
| 136.15 | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 8487 | |
| CC(=O)C1=CC(=CC=C1)O |
| C8H8O2 | |
| MFCD00002298 | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| 1-(3-hydroxyphenyl)ethanone |
Specifications
| 121-71-1 | |
| 153°C | |
| MFCD00002298 | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| CC(=O)C1=CC(=CC=C1)O | |
| 136.15 | |
| 136.15 | |
| Crystalline Powder |
| 94°C | |
| C8H8O2 | |
| 25 g | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 1-(3-hydroxyphenyl)ethanone | |
| 8487 | |
| ≥98.0% (GC,T) | |
| 3′-Hydroxyacetophenone |
Safety and Handling
EINECSNumber : (3)-1240
TSCA : Yes