Learn More
3'-Hydroxyacetophenone, 99+%
CAS: 121-71-1 | C8H8O2 | 136.15 g/mol
$36.92 - $36.92
Chemical Identifiers
| CAS | 121-71-1 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00002298 |
| InChI Key | LUJMEECXHPYQOF-UHFFFAOYSA-N |
| Synonym | 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol |
| PubChem CID | 8487 |
| IUPAC Name | 1-(3-hydroxyphenyl)ethanone |
| SMILES | CC(=O)C1=CC(=CC=C1)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC411910050
|
Thermo Scientific Chemicals
411910050 |
5 g | Glass bottle |
Each for $36.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 121-71-1 | |
| 136.15 | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 8487 | |
| CC(=O)C1=CC(=CC=C1)O |
| C8H8O2 | |
| MFCD00002298 | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| 1-(3-hydroxyphenyl)ethanone |
Specifications
| 121-71-1 | |
| 100.0 | |
| Beige to Brown | |
| Authentic | |
| Glass bottle | |
| MFCD00002298 | |
| 08, 86 | |
| LUJMEECXHPYQOF-UHFFFAOYSA-N | |
| 1-(3-hydroxyphenyl)ethanone | |
| 8487 | |
| 99+% | |
| 3'-Hydroxyacetophenone, 99+% |
| 99.0 | |
| 94.0°C to 97.0°C | |
| 296.0°C | |
| 99% min. (GC) | |
| C8H8O2 | |
| 5 g | |
| 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol | |
| CC(=O)C1=CC(=CC=C1)O | |
| 136.15 | |
| 136.15 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously
GHS Signal Word: Warning
EINECSNumber : 204-494-0
TSCA : TSCA
RUO – Research Use Only