missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetophenone, 99+%

Catalog No. AC411910050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411910050 5 g Glass bottle
AC411911000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411910050 Supplier Thermo Scientific Chemicals Supplier No. 411910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-71-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-71-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002298
InChI Key LUJMEECXHPYQOF-UHFFFAOYSA-N
Synonym 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
PubChem CID 8487
IUPAC Name 1-(3-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 94.0°C to 97.0°C
Color Beige to Brown
Boiling Point 296.0°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 08, 86
Formula Weight 136.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3'-Hydroxyacetophenone, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously
EINECSNumber 204-494-0
TSCA TSCA

RUO – Research Use Only