missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetophenone, 98%

Catalog No. AAA1254722
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1254722 100 g
AAA1254736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1254722 Supplier Thermo Scientific Chemicals Supplier No. A1254722

CAS: 121-71-1 | C8H8O2 | 136.15 g/mol

3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3’Hydroxyacetophenone is used in the preparation of chalcones and flavonoids, which finds application as an anti-tuberculosis agent and antileishmanial chaocones. It is also involved in the preparation of phenylephrine. Further, it is an active component of castoreum, which is used as a tincture in some perfumes and as a food additive.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and bases.

Chemical Identifiers

CAS 121-71-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002298
InChI Key LUJMEECXHPYQOF-UHFFFAOYSA-N
Synonym 3'-hydroxyacetophenone, 3-hydroxyacetophenone, 1-3-hydroxyphenyl ethanone, m-hydroxyacetophenone, 3-acetylphenol, 1-3-hydroxyphenyl ethan-1-one, ethanone, 1-3-hydroxyphenyl, m-acetylphenol, 3-hydroxy acetophenone, 3-acetophenol
PubChem CID 8487
IUPAC Name 1-(3-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)O

Specifications

Melting Point 93°C to 97°C
Density 1.1
Boiling Point 296°C
Quantity 100 g
Beilstein 2040676
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 3'-Hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-494-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only