missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyacetanilide 98.0+%, TCI America™

Catalog No. H018825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H018825G 25 g
H0188500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H018825G Supplier TCI America Supplier No. H018825G

Chemical Identifiers

CAS 621-42-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002263
InChI Key QLNWXBAGRTUKKI-UHFFFAOYSA-N
Synonym 3-acetamidophenol, n-3-hydroxyphenyl acetamide, metacetamol, 3'-hydroxyacetanilide, m-acetamidophenol, 3-hydroxyacetanilide, n-acetyl-m-aminophenol, m-hydroxyacetanilide, 3-acetylamino phenol, metalid
PubChem CID 12124
ChEBI CHEBI:76987
IUPAC Name N-(3-hydroxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)O

Specifications

Melting Point 148°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Hydroxyacetanilide
EINECSNumber (3)-0678
RTECSNumber AE4100000
TSCA Yes