missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-N-(2-naphthyl)-2-naphthamide 97.0+%, TCI America™

Catalog No. H031925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H031925G Supplier TCI America Supplier No. H031925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-64-8
Molecular Formula C21H15NO2
Molecular Weight (g/mol) 313.36
MDL Number MFCD00053488
InChI Key PMYDPQQPEAYXKD-UHFFFAOYSA-N
Synonym 2-Hydroxy-3-naphthoic Acid 2-Naphthylamide, Naphthol AS-SW, Azoic Coupling Component 7
PubChem CID 67276
IUPAC Name 3-hydroxy-N-(naphthalen-2-yl)naphthalene-2-carboxamide
SMILES OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=C2C=CC=CC2=C1

Specifications

Quantity 25 g
Formula Weight 313.36
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-N-(2-naphthyl)-2-naphthamide
EINECSNumber (5)-2276
TSCA Yes