missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-alpha-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. H14335G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14335G 5 g
H14331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14335G Supplier TCI America Supplier No. H14335G
Only null left

Chemical Identifiers

CAS 2415-09-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00238621
InChI Key COJRWHSKVYUZHQ-UHFFFAOYSA-N
Synonym 1-(3-Hydroxyphenyl)ethanol, 3-(1-Hydroxyethyl)phenol
PubChem CID 13542886
IUPAC Name 3-(1-hydroxyethyl)phenol
SMILES CC(C1=CC(=CC=C1)O)O

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-alpha-methylbenzyl Alcohol