missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-7-methoxy-2-naphthoic Acid 97.0+%, TCI America™

Catalog No. p-7130682
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07075G 5 g
H070725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07075G Supplier TCI America Supplier No. H07075G

Chemical Identifiers

CAS 143355-56-0
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00059082
InChI Key UDAQUPSJOGVPIX-UHFFFAOYSA-N
Synonym 3-Hydroxy-7-methoxy-2-naphthalenecarboxylic Acid
PubChem CID 608330
IUPAC Name 3-hydroxy-7-methoxynaphthalene-2-carboxylic acid
SMILES COC1=CC2=CC(=C(C=C2C=C1)O)C(=O)O

Specifications

Quantity 5 g
Formula Weight 218.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-7-methoxy-2-naphthoic Acid