missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-methylpyridine, 99%

Numéro de catalogue. AC148110250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC148110250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC148110250 Fournisseur Thermo Scientific Chemicals Code fournisseur 148110250
Il en reste null

CAS: 1121-78-4 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1121-78-4
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006339
InChI Key DHLUJPLHLZJUBW-UHFFFAOYSA-N
Synonym 5-hydroxy-2-methylpyridine, 2-methyl-5-hydroxypyridine, 3-hydroxy-6-methylpyridine, 6-methyl-3-pyridinol, 3-pyridinol, 6-methyl, 6-methyl-3-hydroxypyridine, 5-hydroxy-2-picoline, unii-uke16jp1jr, 2-methyl-5-pyridinol, uke16jp1jr
PubChem CID 14275
IUPAC Name 6-methylpyridin-3-ol
SMILES CC1=NC=C(C=C1)O

Spécifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 168°C to 171°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C5H4N)CH2OH
Quantity 25 g
Beilstein 21, III, 480
Formula Weight 109.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-6-methylpyridine, 99%
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 214-337-8
RTECSNumber UU7714900
TSCA TSCA

RUO – Research Use Only