accessibility menu, dialog, popup

UserName

CH3OHN

CAS RN 1121-78-4

IUPAC Name: 6-methylpyridin-3-ol
Synonyms: 3-pyridinol, 6-methyl-
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: DHLUJPLHLZJUBW-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)