missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylisoxazole 98.0+%, TCI America™

Catalog No. p-7130996
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
H13485G 5 g
H134825G 25 g
2 options

Catalog No. H13485G

Supplier: TCI America H13485G

Only null left

Chemical Identifiers

CAS 10004-44-1
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00144468
InChI Key KGVPNLBXJKTABS-UHFFFAOYSA-N
Synonym hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
PubChem CID 24781
ChEBI CHEBI:5827
IUPAC Name 5-methyl-1,2-oxazol-3-one
SMILES CC1=CC(=O)NO1

Specifications

Melting Point 86°C
Quantity 5 g
Formula Weight 99.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-5-methylisoxazole
RTECSNumber NY2932000
TSCA No
Recommended Storage Freezer