accessibility menu, dialog, popup

UserName

CAS RN 10004-44-1

IUPAC Name: 5-methyl-2,3-dihydro-1,2-oxazol-3-one
Synonyms: 5-methyl-3(2H)-isoxazolone 3-isoxazolone,5-methyl
Molecular Weight (g/mol): 99.09
Molecular Formula: C4H5NO2
InChi Key: KGVPNLBXJKTABS-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NO1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)