missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-5-methylisoxazole, 97%

Catalog No. p-7059348
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4445006 5 g
AA4445014 25 g
AA4445022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4445006 Supplier Thermo Scientific Chemicals Supplier No. 04445006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10004-44-1 | C4H5NO2 | 99.089 g/mol

3-Hydroxy-5-methylisoxazole is used in plant growth stimulation activity and seedlings growing. And it is also used in DNA/RNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10004-44-1
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00144468
InChI Key KGVPNLBXJKTABS-UHFFFAOYSA-N
Synonym hymexazol, 3-hydroxy-5-methylisoxazole, hymexazole, 5-methylisoxazol-3-ol, 5-methylisoxazol-3 2h-one, hydroxyisoxazole, tachigaren, itachigarden, 5-methyl-3-hydroxyisoxazole, 5-methyl-3 2h-isoxazolone
PubChem CID 24781
ChEBI CHEBI:5827
IUPAC Name 5-methyl-1,2-oxazol-3-one
SMILES CC1=CC(=O)NO1

Specifications

Quantity 5 g
Merck Index 14,4856
Solubility Information Soluble in alcohol, acetone, THF, chloroform
Formula Weight 99.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Hydroxy-5-methylisoxazole
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
EINECSNumber 233-000-6
RTECSNumber NY2932000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only