missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-4-nitrobenzoic Acid 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 183.12 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Hydroxy-4-nitrobenzoic Acid |
Safety and Handling
| TSCA | No |