missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzoic acid, 98%

CAS: 619-14-7 | C7H5NO5 | 183.12 g/mol

$417.33 - $417.33

Chemical Identifiers

CAS 619-14-7
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007110
InChI Key XLDLRRGZWIEEHT-UHFFFAOYSA-N
Synonym 3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
PubChem CID 69265
IUPAC Name 3-hydroxy-4-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC121671000
SDS View Documents
Thermo Scientific Chemicals
121671000
100 g Plastic bottle
Each for $417.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-14-7
183.12
XLDLRRGZWIEEHT-UHFFFAOYSA-N
69265
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
C7H5NO5
MFCD00007110
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
3-hydroxy-4-nitrobenzoic acid

Specifications

619-14-7
100.0
Brown to Yellow
98%
C7H5NO5
MFCD00007110
10, 146
Solubility in water: insoluble
OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
183.12
183.12
Powder
97.5
233°C
Authentic
Plastic bottle
HOC6H3(NO2)CO2H
100 g
3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
XLDLRRGZWIEEHT-UHFFFAOYSA-N
3-hydroxy-4-nitrobenzoic acid
69265
98%
3-Hydroxy-4-nitrobenzoic acid, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 210-580-9

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only