missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-nitrobenzoic acid, 98%

Catalog No. AAA1072406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1072406 5 g
AAA1072414 25 g
AAA1072422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1072406 Supplier Thermo Scientific Chemicals Supplier No. A1072406

CAS: 619-14-7 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-14-7
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007110
InChI Key XLDLRRGZWIEEHT-UHFFFAOYSA-N
Synonym 3-hydroxy-4-nitrobenzoic acid, 3-hydroxy-4-nitro-benzoic acid, 4-nitro-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-4-nitro, 3-hydroxy-4-nitrobenzoicacid, pubchem17135, acmc-209mxi, 3-hydroxy4nitrobenzoic acid, ksc356q8j, 3-hydroxy-4-nitro benzoic acid
PubChem CID 69265
IUPAC Name 3-hydroxy-4-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O

Specifications

Melting Point 232°C to 236°C
Quantity 5 g
Beilstein 2107564
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-580-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only