missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxybenzoic acid, 97%

Catalog No. p-7021395
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1370906 5 g
AAA1370914 25 g
AAA1370922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1370906 Supplier Thermo Scientific Chemicals Supplier No. A1370906

CAS: 645-08-9 | C8H8O4 | 168.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-08-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002507
InChI Key LBKFGYZQBSGRHY-UHFFFAOYSA-N
Synonym isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol
PubChem CID 12575
ChEBI CHEBI:63798
IUPAC Name 3-hydroxy-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)O

Specifications

Melting Point 251°C to 255°C
Quantity 5 g
Beilstein 2208365
Formula Weight 168.15
Percent Purity ≥97%
Chemical Name or Material 3-Hydroxy-4-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-430-5
RTECSNumber BZ4850000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only