missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. H14965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14965G 5 g
H14961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14965G Supplier TCI America Supplier No. H14965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58123-77-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.02
MDL Number MFCD02068386
InChI Key UABBBWVTEWIIMN-UHFFFAOYSA-N
Synonym 3-hydroxy-4-iodo-benzoic acid, benzoic acid, 3-hydroxy-4-iodo, acmc-1ay71, ksc497o7h, 4-iodo-3-hydroxybenzoic acid, 4-iodo-3-hydroxy-benzoic acid, 3-hydroxy-4-iodobenzoic acid, 4-iodo-3-hydroxy benzoic acid
PubChem CID 736854
IUPAC Name 3-hydroxy-4-iodobenzoic acid
SMILES OC(=O)C1=CC(O)=C(I)C=C1

Specifications

Melting Point 226°C
Color White
Quantity 5 g
Formula Weight 264.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-iodobenzoic Acid