missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methylglutaric Acid 95.0+%, TCI America™

Catalog No. H0436100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0436100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0436100MG Supplier TCI America Supplier No. H0436100MG
Only null left

Chemical Identifiers

CAS 503-49-1
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00002712
InChI Key NPOAOTPXWNWTSH-UHFFFAOYSA-N
PubChem CID 1662
ChEBI CHEBI:16831
IUPAC Name 3-hydroxy-3-methylpentanedioic acid
SMILES CC(CC(=O)O)(CC(=O)O)O

Specifications

Melting Point 109°C
Color White-Yellow
Boiling Point 120°C
Quantity 100 mg
Formula Weight 162.14
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-3-methylglutaric Acid
RTECSNumber MA3753000
TSCA No
Recommended Storage Refrigerator