accessibility menu, dialog, popup

UserName

CAS RN 503-49-1

H3COHHOOOHO

CAS RN 503-49-1

IUPAC Name: 3-hydroxy-3-methylpentanedioic acid
Synonyms: meglutol
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: NPOAOTPXWNWTSH-UHFFFAOYSA-N
SMILES: CC(O)(CC(O)=O)CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results